missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Androgen Receptor (pSer216) Rabbit Polyclonal Antibody with BSA and azide

Stain formalin-fixed, paraffin-embedded tissues with Epredia™ Androgen Receptor, Rabbit Polyclonal Antibody.

Brand:  Epredia™ RB-9030-P

 View more versions of this product

Product Code. 12683357

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The androgen receptor (AR) is a member of the steroid superfamily of ligand-dependent transcription factors. The expression of AR is reportedly inversely correlated with histologic grade, i.e. well-differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminal

Immunogen: A synthetic peptide derived from N-terminal of rat AR

Molecular Weight: 110kDa

Positive Control: LnCAp cells or prostate CA

Cellular Localization: Nuclear

This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
Specifications

Specifications

Androgen Receptor
Polyclonal
200µg/mL
Rabbit
1 mL
Oncogenesis
Human, Mouse
IgG
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Polyclonal
Unconjugated
A synthetic peptide derived from N-terminal of rat AR
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers