Learn More
Thermo Scientific Chemicals L-Threonine, Cell Culture Reagent
68.14 € - 215.07 €
Chemical Identifiers
CAS | 72-19-5 |
---|---|
Molecular Formula | C4H9NO3 |
Molecular Weight (g/mol) | 119.12 |
MDL Number | MFCD00064270 |
InChI Key | AYFVYJQAPQTCCC-UHFFFAOYNA-N |
Synonym | l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin |
PubChem CID | 6288 |
ChEBI | CHEBI:16857 |
IUPAC Name | (2S,3R)-2-amino-3-hydroxybutanoic acid |
SMILES | CC(O)C(N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15495889
|
Thermo Scientific Chemicals
J63709.30 |
250 g |
68.14 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405899
|
Thermo Scientific Chemicals
J63709.A1 |
1 kg |
215.07 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Threonine is used to treat various nervous system disorders including spinal spasticity, multiple sclerosis, familial spastic paraparesis, and amyotrophic lateral sclerosis (ALS, Lou Gehrig's disease). Useful with intestinal disorders and indigestion, threonine has also been used to alleviate anxiety and mild depression. . L-threonine has been used clinically with the aim of increasing glycine concentrations in the cerebral spinal fluid of patients with spasticity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water 50 mg/mL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
72-19-5 | |
119.12 | |
AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
6288 | |
(2S,3R)-2-amino-3-hydroxybutanoic acid |
C4H9NO3 | |
MFCD00064270 | |
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin | |
CHEBI:16857 | |
CC(O)C(N)C(O)=O |
Specifications
256°C (decomposition) | |
White | |
MFCD00064270 | |
14,9380 | |
Soluble in water 50mg/mL. | |
CC(O)C(N)C(O)=O | |
119.12 | |
CHEBI:16857 | |
Cell Culture Reagent | |
L-Threonine |
72-19-5 | |
C4H9NO3 | |
1721646 | |
l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin | |
AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
(2S,3R)-2-amino-3-hydroxybutanoic acid | |
6288 | |
119.12 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 200-774-1
missing translation for 'rtecsNumber' : XO8590000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only