missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Pantolactone, 98%, Thermo Scientific™

Product Code. 15412178
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-(+)-Pantolactone, is used as Pharmaceutical material and intermediate. It is used as an Intermediate in organic syntheses.

Solubility
Soluble in water, hardly soluble in alcohol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Heat sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5405-40-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00216625
InChI Key SERHXTVXHNVDKA-SCSAIBSYSA-N
Synonym s-+-pantolactone, l-pantolactone, s-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, s-pantolactone, l-+-pantoyl lactone, 3s-3-hydroxy-4,4-dimethyloxolan-2-one, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3s, pantolactons, l-pantoyl lactone
PubChem CID 736053
IUPAC Name (3S)-3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 89°C to 94°C
Boiling Point 120°C to 122°C (15 mmHg)
Quantity 1 g
Beilstein 4659425
Solubility Information Soluble in water,hardly soluble in alcohol.
Optical Rotation +51° (c=3 in Water)
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material L-(+)-Pantolactone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.