missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, 99%, Thermo Scientific Chemicals

Product Code. 11472277
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture.

L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.

Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in methanol, ether, benzene, acetone, ethyl acetate and chloroform.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185°C (decomposition)
Beilstein 1723797
Packaging Glass Bottle
Solubility Information Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in methanol,ether,benzene,acetone,ethyl acetate and chloroform.
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
Formula Weight 146.15
Percent Purity 99%
Quantity 1000 g
Physical Form Powder
Chemical Name or Material L-Glutamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'