missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, 99%, Thermo Scientific Chemicals

Product Code. 10619094
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle

L-Cystine, CAS # 56-89-3 is the left-handed enantiomer of the sulphur-containing amino acid, cystine, purified for use in molecular biology and analytical chemistry applications.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • L-Cystine is a derivative of the amino acid, cysteine
  • Cystine is not considered one of the twenty common amino acids, but it has a role as a metabolite in some species
  • May act as an antioxidant in form and reduce aging, inflammation, and tissue damage

Applications

  • Used as an additive to cell culture media
  • Intended for biochemistry applications such as HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
MDL Number MFCD00064228
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
PubChem CID 67678
ChEBI CHEBI:16283
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Ammonium 200ppm max.
Color White to Beige
Chloride 200ppm max.
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula [-SCH2CH(NH2)CO2H]2
Iron (Fe) 10ppm max.
Beilstein 04, 507
Loss on Drying 0.5 % max. (105°C, 3 hrs)
Merck Index 15, 2779
Packaging Plastic Bottle
Specific Rotation −209° to −224° (20°C, 589nm) (c=2, 1 M HCl) on dried substance
Sulfate 300ppm max.
Formula Weight 240.3
Percent Purity ≥98.5% (Acid)
Sulfated Ash 0.1 % max.
Specific Rotation Condition − 224.00 (20.00°C c=2;1 M HCL dry.sub.)
Quantity 500 g
Physical Form Fused Solid, Crystals or Chunks
Chemical Name or Material L-Cystine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'