missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-beta-Proline, 98+%, Thermo Scientific Chemicals
102.00 € - 343.00 €
Chemical Identifiers
CAS | 72580-53-1 |
---|---|
Molecular Formula | C5H9NO2 |
Molecular Weight (g/mol) | 115.132 |
MDL Number | MFCD06738463 |
InChI Key | JAEIBKXSIXOLOL-BYPYZUCNSA-N |
Synonym | s-pyrrolidine-3-carboxylic acid, s-+-pyrrolidine-3-carboxylic acid, l-beta-proline, 3s-3-pyrrolidinecarboxylic acid, 3s-pyrrolidine-3-carboxylic acid, 3-pyrrolidinecarboxylic acid, 3s, s-pyrrolidine-3-carboxylicacid, s-beta-proline, s-, a-proline, pubchem11126 |
PubChem CID | 1501969 |
IUPAC Name | (3S)-pyrrolidine-3-carboxylic acid |
SMILES | C1CNCC1C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15464288
|
Thermo Scientific Alfa Aesar
H57895.MD |
250 mg |
102.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15454288
|
Thermo Scientific Alfa Aesar
H57895.03 |
1 g |
343.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an agrochemical and pharmacetical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an agrochemical and pharmacetical intermediate.
Solubility
Slightly soluble in water.
Notes
Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents, acids.
Chemical Identifiers
72580-53-1 | |
115.132 | |
JAEIBKXSIXOLOL-BYPYZUCNSA-N | |
1501969 | |
C1CNCC1C(=O)O |
C5H9NO2 | |
MFCD06738463 | |
s-pyrrolidine-3-carboxylic acid, s-+-pyrrolidine-3-carboxylic acid, l-beta-proline, 3s-3-pyrrolidinecarboxylic acid, 3s-pyrrolidine-3-carboxylic acid, 3-pyrrolidinecarboxylic acid, 3s, s-pyrrolidine-3-carboxylicacid, s-beta-proline, s-, a-proline, pubchem11126 | |
(3S)-pyrrolidine-3-carboxylic acid |
Specifications
185°C to 191°C | |
C5H9NO2 | |
3537293 | |
Slightly soluble in water. | |
C1CNCC1C(=O)O | |
115.132 | |
115.13 | |
+18° (c=2 in Water) |
72580-53-1 | |
MFCD06738463 | |
s-pyrrolidine-3-carboxylic acid, s-+-pyrrolidine-3-carboxylic acid, l-beta-proline, 3s-3-pyrrolidinecarboxylic acid, 3s-pyrrolidine-3-carboxylic acid, 3-pyrrolidinecarboxylic acid, 3s, s-pyrrolidine-3-carboxylicacid, s-beta-proline, s-, a-proline, pubchem11126 | |
JAEIBKXSIXOLOL-BYPYZUCNSA-N | |
(3S)-pyrrolidine-3-carboxylic acid | |
1501969 | |
≥98% | |
L-beta-Proline |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only