missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kryptofix(R) 222, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 68.0°C to 72.0°C |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Quantity | 5 g |
Merck Index | 15,2596 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,diethylether and,chloroform |
Formula Weight | 376.48 |
Percent Purity | 98% |
Chemical Name or Material | Kryptofix* 222 |
Safety and Handling
Product Identifier
- Kryptofix 222
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Kryptofix 222
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'