Learn More
DWK Life Sciences Kimble™ Kontes™ Hollow White Rubber Sleeve Stoppers
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
Brand:  DWK Life Sciences 774261-0024
 View more versions of this product
Product Code. 12869518
  • 212.00 € / Pack of 50
Estimated Shipment: 19-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Hollow stopper fits into the neck of the glassware apparatus. Series 774261 may be used at up to 125°C for up to 8 hours.
  • Plug-type stopper with sleeve extension fits into the neck of the apparatus for a secure seal
  • Diaphragm can be punctured with a syringe needle
  • Use for outgassing NMR sample tubes, as a pressure vent or for adding reactant via a syringe
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2.4 cm
24/40; 24/20
Rubber
NMR tubes, pressure venting or adding reactants via syringe
White
Plug-type Sleeve Stopper