Learn More
Kojic acid, 99%, Thermo Scientific Chemicals
43.72 € - 458.28 €
Chemical Identifiers
CAS | 501-30-4 |
---|---|
Molecular Formula | C6H6O4 |
Molecular Weight (g/mol) | 142.11 |
MDL Number | MFCD00006580 |
InChI Key | BEJNERDRQOWKJM-UHFFFAOYSA-N |
Synonym | kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52 |
PubChem CID | 3840 |
ChEBI | CHEBI:43572 |
IUPAC Name | 5-hydroxy-2-(hydroxymethyl)pyran-4-one |
SMILES | OCC1=CC(=O)C(O)=CO1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439652
|
Thermo Scientific Alfa Aesar
A10760.06 |
5 g |
43.72 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449652
|
Thermo Scientific Alfa Aesar
A10760.14 |
25 g |
147.74 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459652
|
Thermo Scientific Alfa Aesar
A10760.22 |
100 g |
458.28 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kojic acid may be used on cut fruits to prevent oxidative browning, in seafood to preserve pink and red colors and in cosmetics to lighten skin. It is used to treat skin diseases like melasma as it has antibacterial and antifungal properties. It is used as a chelation agent that inhibits melanin production and also as a tyrosine kinase inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water, acetone (freely soluble), ethanol (freely soluble), methanol, DMSO, ethyl acetate (slightly soluble), chloroform (slightly soluble), pydidine (slightly soluble) and ether (slightly soluble). Insoluble in benzene.
Notes
Should be stored at room temperature. Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Chemical Identifiers
501-30-4 | |
142.11 | |
BEJNERDRQOWKJM-UHFFFAOYSA-N | |
3840 | |
5-hydroxy-2-(hydroxymethyl)pyran-4-one |
C6H6O4 | |
MFCD00006580 | |
kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52 | |
CHEBI:43572 | |
OCC1=CC(=O)C(O)=CO1 |
Specifications
501-30-4 | |
Odorless | |
MFCD00006580 | |
14,5317 | |
Soluble in water,acetone (freely soluble),ethanol (freely soluble),methanol,DMSO,ethyl acetate (slightly soluble),chloroform (slightly soluble),pydidine (slightly soluble) and ether (slightly soluble). Insoluble in benzene. | |
OCC1=CC(=O)C(O)=CO1 | |
142.11 | |
CHEBI:43572 | |
99% |
152°C to 155°C | |
C6H6O4 | |
120895 | |
kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52 | |
BEJNERDRQOWKJM-UHFFFAOYSA-N | |
5-hydroxy-2-(hydroxymethyl)pyran-4-one | |
3840 | |
142.11 | |
Kojic acid |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P280-P308+P313-P501c
H351
missing translation for 'einecsNumber' : 207-922-4
missing translation for 'rtecsNumber' : UQ0875000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.