missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17117-1-AP-150UL
This item is not returnable.
View return policy
Description
Kelch-like protein 11, Subcellular location Secreted.Specifications
KLHL11 | |
Polyclonal | |
Unconjugated | |
KLHL11 | |
kelch like 11 (Drosophila), Kelch like protein 11, KLHL11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217194, 287706, 55175 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CE33, Q9NVR0 | |
Klhl11 | |
KLHL11 Fusion Protein Ag10805 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction