missing translation for 'onlineSavingsMsg'
Learn More

Kinetin, 99%, Thermo Scientific Chemicals

65.12 € - 534.66 €

Chemical Identifiers

CAS 525-79-1
Molecular Formula C10H9N5O
Molecular Weight (g/mol) 215.21
InChI Key QANMHLXAZMSUEX-UHFFFAOYSA-N
Synonym kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
PubChem CID 3830
ChEBI CHEBI:27407
IUPAC Name N-(furan-2-ylmethyl)-7H-purin-6-amine
SMILES C1=COC(=C1)CNC2=NC=NC3=C2NC=N3
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10698381
View Documents
Thermo Scientific Acros
226500010
1 g Glass bottle
65.12 €
1g
Please to purchase this item. Need a web account? Register with us today!
10771901
View Documents
Thermo Scientific Acros
226500050
5 g Glass bottle
219.09 €
5g
Please to purchase this item. Need a web account? Register with us today!
11996811
View Documents
Thermo Scientific Acros
226500250
25 g Glass bottle
534.66 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

525-79-1
215.21
kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
CHEBI:27407
C1=COC(=C1)CNC2=NC=NC3=C2NC=N3
C10H9N5O
QANMHLXAZMSUEX-UHFFFAOYSA-N
3830
N-(furan-2-ylmethyl)-7H-purin-6-amine

Specifications

525-79-1
White to Beige
99%
26,IV,3586
kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
QANMHLXAZMSUEX-UHFFFAOYSA-N
N-(furan-2-ylmethyl)-7H-purin-6-amine
3830
215.21
Fine Crystalline Powder
269.0°C to 271.0°C
Authentic
C10H9N5O
15,5359
Solubility in water: soluble. Other solubilities: freely soluble in dilute aquous hcl or naoh,slightly soluble in methanol and ethanol
C1=COC(=C1)CNC2=NC=NC3=C2NC=N3
215.21
CHEBI:27407
99%
Kinetin
Videos
Safety and Handling

Safety and Handling

Warning

missing translation for 'einecsNumber' : 208-382-2

Product Identifier
  • Kinetin

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.