Learn More
Kimberly-Clark Professional™ Kimtech Pure™ A6 Clean Overalls
Maintain ISO 7 Class C or higher cleanroom environments with breathable, ultra light overalls. The Kimtech Pure™ A6 Clean Overalls are made of a high performance, chemical resistant fabric that are origami folded to reduce gowning time.
Specifications
Product Type Cleanroom Coveralls
Material SMS
Color White
Disposable Single-use
Cuff Style Thumb Loop
View More Specs
Description
Description
  • High performance chemical resistant fabric
  • Origami folded to reduce gowning time
  • Presented unzipped and inside-out reducing the risk of contaminating the outside of the garment
Specifications
Specifications
Cleanroom Coveralls
White
Thumb Loop
Non-sterile
Breathable
Controlled Environments
SMS
Single-use
Collared
Standing
PPE Cat III Type 5/6 (Protection against particles/limited splash protection). Add Infectious agent protection. EN1149-1:1995 Antistatic garment