Learn More
Kimberly-Clark Professional™ Kimtech™ G5 Sterile Latex Hand Specific Gloves
Kimtech™ G5 Sterile Latex gloves provide sensitive and dextrous protection in EU GMP ISO Class 5 Grade A sterile cleanrooms. The hand-specific gloves are suitable for double-donning and feature a textured finish for enhanced tactile performance.
Specifications
Certifications/Compliance ISO 9001; 0123; Category III PPE - CE Complex
Cleanroom Class Class 100 (ISO 5)
Exterior Finish Fully Textured
Hand Specific
Material Latex
View More Specs
Description
Description
Kimtech™ G5 Sterile Latex gloves provide sensitive and dextrous protection in EU GMP ISO Class 5 Grade A sterile cleanrooms. The hand-specific gloves are suitable for double-donning and feature a textured finish for enhanced tactile performance.
  • Assured Compliance:
    • PPE Cat III according to Regulation (EU) 2016/425
    • EN ISO 374-1:2016 Type C (K) Chemical Splash protection
    • EN 374-4:2003 Resistance to degradation by chemicals
    • EN ISO 374-5:2016 Micro Organism and Virus Protection
    • Sterility Assurance Level (SAL) 10-6
    • Certificate of Analysis available online
    • Certificate of Sterility available online
  • The fully sterilized latex material provides high levels of contamination protection against proteins, particles, micro-organisms, and chemical splash, and low endotoxin levels.
  • Suitable for use in EU GMP ISO Class 5 Grade A or higher sterile cleanrooms, and provided with supporting technical information to demonstrate regulatory compliance.
  • Gloves are hand-specific, disposable and a natural color, with a high level of cleanliness.
  • Textured palm and fingertips enhance grip and tactile sensitivity for safer and more efficient processes.
  • Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
  • Made from latex sterilized with gamma radiation and containing no powder, reducing the risks of skin irritation for the wearer.
Specifications
Specifications
ISO 9001; 0123; Category III PPE - CE Complex
Fully Textured
Latex
Beaded
30 cm
Class 100 (ISO 5)
Specific
Sterile
28 Mpa (27 Mpa after aging)