Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Autoclavable and used for multiple injections
Description
- Manufactured using the highest quality raw materials
- Designed for an ideal fit with Kimble Chase autosampler vials
- Convenient assembly of caps and liners
- Available with standard or pre-slit septa
Specifications
Polypropylene | |
Autoclavable |