Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Pipets
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
Specifications
Material Borosilicate Glass
Quantity 100/Cs.
View More Specs
Description
Description
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
  • Available with 7 and 9 in. (17.7 and 23cm) tips for use with 7 and 9 in. long NMR tubes
Specifications
Specifications
Borosilicate Glass
100/Cs.