missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Screw-Thread N-51A Borosilicate Glass Sample Vials: Without Caps

Low extractable Kimax N-51A glass

Specifications

Classification B
Color Clear
Material Borosilicate Glass
For Use With (Application) For general purpose
Thread Style Screw
View More Specs

Description

Description

Low extractable Kimax N-51A glass

  • Borosilicate glass sample vials without caps

Compliance

Kimax N-51A glass conforms to ASTM Type 1, Class B requirements.

Specifications

Specifications

B
Borosilicate Glass
Screw
ASTM Type I
Clear
For general purpose
Screw Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Kimble™ Screw-Thread N-51A Borosilicate Glass Sample Vials: Without Caps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.