missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps
Packaged in corrugated trays with partitions
Specifications
Classification | A |
---|---|
Material (Liner) | PTFE/Rubber |
Color | Clear |
Material | Borosilicate Glass |
Thread Style | Screw |
Description
- Small case quantities
- Manufactured from clear 33 expansion, borosilicate glass
- Attached closed top black phenolic closures
Specifications
A | |
Clear | |
Screw |
PTFE/Rubber | |
Borosilicate Glass | |
Phenolic |