missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ G60 Purple Nitrile™ Cut-Resistant Gloves

Product Code. 11803490
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Cut-resistant gloves are constructed of comfortable, breathable fabric and feature purple tapered coating on the fingertips for added dexterity.

High level of cut resistance while enhancing dexterity, grip and comfort.
  • Dyneema™ fabric construction
  • Purple Nitrile coated fingertips
  • Flexible dotted palm
  • Ambidextrous

Specifications

Coating Material Nitrile Dots
Color White, Purple
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Latex Free Yes
Material Dyneema
Product Type Cut-resistant Gloves
Size 10
EN388 Standard EN 388
For Use With (Application) Glass/Metal handling, Paper making and automotive/aircraft assembly
Glove Cuff Style Knit
Model 97433
Series Kleenguard™ G60
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.