Learn More
Kimberly-Clark Professional™ KleenGuard™ G40 Black Polyurethane Coated Nylon Gloves
Multi-purpose protective gloves designed for tasks that require high dexterity and fine motor skills. The thin-mil design provides tactile sensitivity, making KleenGuard™ G40 Black Polyurethane Coated Nylon Gloves ideal for performing delicate tasks such as electronics assembly.
216.96 €
Specifications
Coated Area Palm, Fingers
Coating Material Polyurethane
Color Black
Exterior Finish Partially Textured
Hand Specific
View More Specs
Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15324252
View Documents
Kimberly-Clark Professional™
13837
Small
216.96 €
60 pairs
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
15334252
View Documents
Kimberly-Clark Professional™
13838
Medium
216.96 €
60 pairs
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
15344252
View Documents
Kimberly-Clark Professional™
13839
Large
216.96 €
60 pairs
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
15354252
View Documents
Kimberly-Clark Professional™
13840
X-Large
216.96 €
60 pairs
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
15364252
View Documents
Kimberly-Clark Professional™
13841
2X-Large
216.96 €
60 pairs
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
Description
Description
  • General purpose protective gloves designed for tasks that require high dexterity and fine motor skills
  • Thin mil design provides tactile sensitivity, ideal for performing delicate tasks
  • Seamless knit nylon backing enhances durability, and provides resistance to tears and abrasion
  • Dark color is ideal for hiding dirt
Specifications
Specifications
Palm, Fingers
Black
Specific
Seamless Knit Gloves
Small Parts Handling, Precision Assembly, and Inspection
G40
Polyurethane
Partially Textured
Nylon
Full-hand Mitt
Knit