missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves

Product Code. 15450490
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves are constructed of ultra-high molecular weight polyethylene fibers to enhance breathability, and provide ANSI Cut Level 3 protection.

  • Constructed of ultra-high molecular weight polyethylene (UHMWPE) fibers to provide breathability and protection
  • Polyurethane coated palm and fingers enhance dexterity and grip capabilities
  • Separate right and left gloves in a pair provide a snug fit

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388
For Use With (Application) Assembly, plastics, light raw materials
Glove Cuff Style Knit
Model 13826
Series KleenGuard™ G60
Yarn Gauge 13
Length (English) 10.2 in.
Length (Metric) 26 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.