Learn More
DWK Life Sciences Kimble™ KIMAX™ Long, Straight-Neck Flask with Capacity Scale
Neck accepts metal or plastic cap or cotton plug
492.00 €
Specifications
Material Borosilicate Glass
For Use With (Application) Metal cap, Plastic closure, Foam plug, or Cotton plug for Cell Culture applications
Neck Style Plain fire-polished
Autoclavable Autoclavable
View More Specs
Products 1
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10389924
View Documents
DWK Life Sciences
25615250
250 mL
492.00 €
Pack of 24
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
Description
Description
  • Thick-walled tubing neck and plain fire-polished finish sealed to an Erlenmeyer-shaped body
  • Graduated to capacity
  • Durable white ceramic enamel scale
Compliance
Designed from ASTM™ E 1404, Type IV, Class 1 requirements
Specifications
Specifications
Borosilicate Glass
Metal cap, Plastic closure, Foam plug, or Cotton plug for Cell Culture applications
Plain fire-polished
Autoclavable