Learn More
DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Serological Pipets with Tooled Opening
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Specifications
Certifications/Compliance | ASTM E1044 |
---|---|
Material | Borosilicate Glass |
Graduated | Yes |
Type | Serological Pipet |
Description
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Order Info
Plugs not included.
Compliance
Calibrated To Deliver/Blow Out. Designed from ASTM™ E 1044.
Specifications
ASTM E1044 | |
Borosilicate Glass | |
Yes | |
Serological Pipet |