missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13403-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of kinesin-like protein family. This family of proteins are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. The C-terminal half of this protein has been shown to bind DNA. Studies with the Xenopus homolog suggests its essential role in metaphase chromosome alignment and maintece.Specifications
KIF22 | |
Polyclonal | |
Unconjugated | |
KIF22 | |
KID, KIF22, kinesin family member 22, Kinesin like protein 4, Kinesin like protein KIF22, KNSL4, OBP, OBP 1, OBP 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110033, 293502, 3835 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14807, Q3V300, Q5I0E8 | |
KIF22 | |
KIF22 Fusion Protein Ag4254 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |