Learn More
KIAA1984 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20945-1-AP-20UL
 View more versions of this product
Product Code. 16810535
  • 145.00 € / 20µL
Explore more special offers
Description
Description
CCDC183 is a protein coding gene.
Specifications
Specifications
KIAA1984
Polyclonal
Unconjugated
CCDC183
KIAA1984, PARF, RP11 216L13.9
Rabbit
Antigen Affinity Chromatography
RUO
84960
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5T5S1
CCDC183
KIAA1984 Fusion Protein Ag15053
20 μL