missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Ki-67, Rabbit Monoclonal Antibody

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Brand:  Epredia™ RM-9106-S0

 View more versions of this product

Product Code. 12683697

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.

Host Species: Rabbit

Clone: SP6

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from the human Ki-67 protein

Positive Control: Spleen, tonsil, or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Ki-67
Monoclonal
Unconjugated
A synthetic peptide derived from the human Ki-67 protein
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
SP6
Rabbit
100 μL
Cancer and Tumor Biology
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers