missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Ki-67, Rabbit Monoclonal Antibody

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Brand:  Epredia™ RM-9106-S1

546.33 EUR valid until 2024-12-31
Use promo code "21882" to get your promotional price.


 View more versions of this product

Product Code. 12603707

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.

Host Species: Rabbit

Clone: SP6

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from the human Ki-67 protein

Positive Control: Spleen, tonsil, or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Ki-67
Monoclonal
Unconjugated
Rabbit
Spleen, tonsil or Breast CA
IVD
Primary
2 to 8°C
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Formalin/Paraffin)
SP6
Ki-67 Monoclonal Antibody, 0.5mL, SP6 Clone, A Synthetic Peptide Derived from the Human Ki-67 Protein Immunogen, Rabbit Host, Human Species Reactivity, Unconjugated, IgG Isotype, C-terminus Epitope, Nuclear Staining Pattern
A synthetic peptide derived from the human Ki-67 protein
500 μL, 0.5mL
Cancer and Tumor Biology
1 Each
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers