Learn More
Ketoconazole, Fisher BioReagents
86.24 € - 915.00 €
Chemical Identifiers
CAS | 65277-42-1 |
---|---|
Molecular Formula | C26H28Cl2N4O4 |
Molecular Weight (g/mol) | 534.452 |
InChI Key | XMAYWYJOQHXEEK-SIULDFEJSA-N |
Synonym | ketoconazole |
PubChem CID | 76973198 |
IUPAC Name | 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone |
SMILES | CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10430015
|
Fisher BioReagents
BP2734-50 |
50 mg | Amber Glass | ||||||
10754344
|
Fisher BioReagents
BP2734-100 |
100 mg | Amber Glass | ||||||
10032624
|
Fisher BioReagents
BP2734-500 |
500 mg | Amber Glass | ||||||
11448353
|
Fisher BioReagents
BP2734-1 |
1 g | Amber Glass | ||||||
Description
Ketoconazole is a cytochrome P450 inhibitor and demonstrates anti-psoriatic activity. An antifungal agent.Chemical Identifiers
65277-42-1 | |
534.452 | |
ketoconazole | |
2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone |
C26H28Cl2N4O4 | |
XMAYWYJOQHXEEK-SIULDFEJSA-N | |
76973198 | |
CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl |
Specifications
65277-42-1 | |
White | |
< 0.5% | |
98% | |
15, 5349 | |
XMAYWYJOQHXEEK-SIULDFEJSA-N | |
2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone | |
76973198 | |
Solid |
148°C | |
< 0.1% | |
Inclusive between 148°C | |
C26H28Cl2N4O4 | |
ketoconazole | |
CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl | |
534.452 | |
531.44 | |
Ketoconazole |
Safety and Handling
WARNING!
Emergency Overview
Toxic if swallowed. May cause skin, eye, and respiratory tract irritation. Danger of serious damage to health by prolonged exposure. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:3
Flammability:0
Instability:0
missing translation for 'dotInformation' : DOT Class 6.1, : Poison
Recommended Storage : RT
- Ketoconazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360F-May damage fertility.
- H373-May cause damage to organs.
- H301-Toxic if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.