missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNE1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13867-1-AP-20UL
This item is not returnable.
View return policy
Description
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a membrane protein which has sequence similarity to the KCNE1 gene product, a member of the potassium channel, voltage-gated, isk-related subfamily. This intronless gene is deleted in AMME contiguous gene syndrome and may be involved in the cardiac and neurologic abnormalities found in the AMME contiguous gene syndrome.Specifications
KCNE1L | |
Polyclonal | |
Unconjugated | |
KCNE5 | |
AMMECR2, KCNE1 like, KCNE1L, KCNE5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23630, 66240, 681190 | |
-20°C | |
Liquid |
Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9QZ26, Q9UJ90 | |
Kcne5 | |
KCNE1L Fusion Protein Ag4849 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction