missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KChIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14212-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. Alternative splicing results in multiple transcript variant encoding different isoforms.Specifications
KChIP1 | |
Polyclonal | |
Unconjugated | |
KCNIP1 | |
KCHIP1, KCNIP1, VABP, Vesicle APC binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
30820 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NZI2 | |
KCNIP1 | |
KChIP1 Fusion Protein Ag5432 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |