missing translation for 'onlineSavingsMsg'
Learn More
Learn More
kazrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11572-1-AP-20UL
This item is not returnable.
View return policy
Description
Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.Specifications
kazrin | |
Polyclonal | |
Unconjugated | |
KAZN | |
KAZ, kazrin, KIAA1026, RP1 21O18.1, RP1-21O18.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23254, 313672, 71529 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5FWS6, Q674X7, Q69ZS8 | |
Kazn | |
kazrin Fusion Protein Ag2152 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction