Learn More
kazrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11572-1-AP-150UL
 View more versions of this product
Product Code. 16848743
  • 410.00 € / 150µL
Explore more special offers
Description
Description
Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.
Specifications
Specifications
kazrin
Polyclonal
Unconjugated
KAZN
KAZ, kazrin, KIAA1026, RP1 21O18.1, RP1-21O18.1
Rabbit
Antigen Affinity Chromatography
RUO
23254, 313672, 71529
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5FWS6, Q674X7, Q69ZS8
Kazn
kazrin Fusion Protein Ag2152
150 μL
Primary
Human, Mouse, Rat