missing translation for 'onlineSavingsMsg'
Learn More

Kaolin, Pure, Light, Washed with Acid, Fisher Chemical™

34.20 € - 86.00 €

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
Synonym kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
PubChem CID 56841936
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10010220
View Documents
Fisher Chemical
K/0050/60
1 kg Plastic powder jar This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10753991
View Documents
Fisher Chemical
K/0050/63
3 kg Plastic keg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1332-58-7
258.16
kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
Al2H4O9Si2
NLYAJNPCOHFWQQ-UHFFFAOYSA-N
56841936

Specifications

1750°C
Beige
Al2H4O9Si2
kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
258.16
258.2g/mol
1332-58-7
Powder/Solid
MFCD00062311
NLYAJNPCOHFWQQ-UHFFFAOYSA-N
oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate
56841936
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Kaolin

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.