missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ K Type Extension Cables
Use these extension cables for a variety of applications. Cole-Parmer™ K Type Extension Cables feature 20-gauge (0.032 in. dia) Thermocouple Wire with PVC/PVC Insulation.
32.95 € - 49.55 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11750979
|
Cole-Parmer™
08516-30 |
3 m |
32.95 €
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||||
11760979
|
Cole-Parmer™
08516-35 |
7.5 m |
49.55 €
Each |
Estimated Shipment: 19-11-2024
Log in to see stock. |
|||||
Description
- Each probe comes with color coded ANSI mini-connectors: type J-black, type K-yellow and type T-blue
Specifications
ANSI | |
Extension Cable |