Learn More
Anritsu™ Type K Magnet Probes
Designed with magnets to allow probes to be attached to measurement targets. Anritsu™ Type K Magnet Probes are temperature sensors designed for stationary surfaces.
Specifications
Analysis Time 2.0 sec.
Type Thermocouple K
Material Chromel-Alumel
View More Specs
Description
Description
  • Designed for stationary surface
  • Available with square or round footprint
  • Magnetic probe that attaches to steel surface
  • Thermocouple contact
Specifications
Specifications
2.0 sec.
Thermocouple K
Chromel-Alumel