missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Sure-Grip™ EX Compac Flammable Safety Cabinet
Justrite Compac Safety Cabinets fit nearly anywhere and provide protection where space is at a premium. The cabinets protect workers, reduce fire risks, and allow storage of flammable fuels and chemicals in code-compliant safety cabinets.
Specifications
Depth (English) Exterior | 18 in. |
---|---|
Depth (Metric) Exterior | 45.7 cm |
Load Bearing Capacity | 12 gal. |
Certifications/Compliance | FM, NFPA, OSHA |
Color | Yellow |
Description
- One-door cabinet stores up to 12 gal. (45.4L) of flammables
- Double-wall construction provides 11/2 in. of insulating air space to help prevent the spread of flames Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- High-visibility Haz-Alert™ reflective warning labels provide added safety during power outages or fires
- Self-latching doors facilitate fire protection of the cabinet
- Self latch engages critical three-point latching system for protection under fire conditions
- Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- One adjustable SpillSlope™ safety shelf directs spills to back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Finished inside and out with a durable lead-free hybrid epoxy and polyester powder paint for chemical resistance
- Fit neatly under counter or stand alone
- Lettering on door reads: “FLAMMABLE—KEEP FIRE AWAY”
- Backed by an exclusive 10-year warranty
- Meet OSHA and NFPA requirements
19-121-852: Manual-close door
19-121-853; Self-closing door
Certifications
FM approved. Self-closing model UL listed.
Compliance
Meet NFPA and OSHA requirements. Self-closing model complies with Uniform Fire Code.
Specifications
18 in. | |
12 gal. | |
Yellow | |
35 in. | |
Storage Cabinet | |
59.05 cm | |
One adjustable shelf, U-Loc™ handle with two keys and accept padlock, Haz-Alert™ reflective exterior labeling and Galvanized Steel SpillSlope™ shelves, leakproof 2 in. bottom sump, dual vents with flam | |
1 | |
Epoxy, Polyester |
45.7 cm | |
FM, NFPA, OSHA | |
14.56 x 20 x 30.5 in. (37 x 50.8 x 77.5 cm) | |
88.9 cm | |
23.25 in. | |
Flammables | |
Steel (CR) | |
1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title