missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Document Storage Box Labels

For use with document storage boxes

Brand:  Justrite™ 23310

Product Code. 12831884

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Includes four yellow, two blank and two printed (SDS and instruction manual) labels

Specifications

Specifications

Storage Box Label
Yellow
Polyethylene
2 blank labels and labels printed SDS and instruction manual
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers