missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Jenway™ Filters
274.00 € - 932.00 €
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
11424859
|
Cole-Parmer™
10089-64 |
245 nm | |||||||
11434849
|
Cole-Parmer™
10089-58 |
320 to 380 nm |
274.00 €
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11484869
|
Cole-Parmer™
10089-65 |
340 nm | |||||||
11414879
|
Cole-Parmer™
10089-70 |
365 nm | |||||||
11444849
|
Cole-Parmer™
99968-74 |
380 to 500 nm |
274.00 €
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11434879
|
Cole-Parmer™
15986-40 |
380 nm |
630.00 €
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11454849
|
Cole-Parmer™
10089-57 |
480 to 580 nm |
274.00 €
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11404899
|
Cole-Parmer™
10089-69 |
514 nm | |||||||
Specifications
Filter | |
Fluorimeter |