missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAML Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | JAML |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890905
|
Proteintech
21302-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880905
|
Proteintech
21302-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMICA transmembrane protein of the plasma membrane of leukocytes that control their migration and activation through interaction with CXADR, a plasma membrane receptor found on adjacent epithelial and endothelial cells. The interaction between both receptors mediates the activation of gamma-delta T-cells, a subpopulation of T-cells residing in epithelia and involved in tissue homeostasis and repair. Upon epithelial CXADR-binding, JAML induces downstream cell signaling events in gamma-delta T-cells through PI3-kinase and MAP kinases. It results in proliferation and production of cytokines and growth factors by T-cells that in turn stimulate epithelial tissues repair. It also controls the transmigration of leukocytes within epithelial and endothelial tissues through adhesive interactions with epithelial and endothelial CXADR.Specifications
JAML | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
Q86YT9 | |
120425 | |
JAML Fusion Protein Ag15900 | |
Primary | |
-20°C | |
JAML |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AMICA, AMICA1, CREA7 1, CREA7 4, Gm638, JAML | |
JAML | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |