missing translation for 'onlineSavingsMsg'
Learn More

GPE Scientific™ J Young Spherical Joint, Female/O-Ring Cup Joint

GPE Scientific™ J Young Spherical Joint, Female/O-Ring Cup Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.

Brand:  GPE Scientific™ GBF/51/35

 View more versions of this product

Product Code. 15874754

  • 54.05 € / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spherical joint designed for use with ball joint
  • Available with different joint sizes, shank bores, and shank ODs
Specifications

Specifications

Shank Bore: 35 mm, Shank OD: 38 mm
Female Spherical Joint
51/35
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers