Learn More
GPE Scientific™ J Young Schlenk Cap
GPE Scientific™ J Young Schlenk Cap is specially designed for use with Schlenk lines and vacuum manifolds.
Brand:  GPE Scientific™ SC/29
 View more versions of this product
Product Code. 15804884
  • 19.95 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cap for Schlenk lines and vacuum manifolds
  • Available in different joint sizes
Specifications
Specifications
J Young Cap
Schlenk lines and vacuum manifolds
Joint size: 29