missing translation for 'onlineSavingsMsg'
Learn More

Isopropylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 15416747
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76347-13-2
Molecular Formula C9H19BO2
Molecular Weight (g/mol) 170.059
MDL Number MFCD05663856
InChI Key MECCSFDHAVAAAW-UHFFFAOYSA-N
Synonym 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 12727711
IUPAC Name 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C(C)C

Specifications

Boiling Point 115°C (60 mmHg)
Refractive Index 1.409
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 170.06
Percent Purity 98%
Chemical Name or Material Isopropylboronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.