Learn More
Isophthaloyl dichloride, 98%, Thermo Scientific Chemicals
27.84 € - 250.00 €
Chemical Identifiers
CAS | 99-63-8 |
---|---|
Molecular Formula | C8H4Cl2O2 |
Molecular Weight (g/mol) | 203.02 |
InChI Key | FDQSRULYDNDXQB-UHFFFAOYSA-N |
Synonym | isophthaloyl chloride, isophthaloyl dichloride, 1,3-benzenedicarbonyl dichloride, m-phthaloyl chloride, isophthalic chloride, isophthalic acid chloride, isophthalic acid dichloride, isothaloyl chloride, isophthalyl chloride, 1,3-benzenedicarbonyl chloride |
PubChem CID | 7451 |
IUPAC Name | benzene-1,3-dicarbonyl chloride |
SMILES | C1=CC(=CC(=C1)C(=O)Cl)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10775381
|
Thermo Scientific Acros
122661000 |
100 g | Glass bottle |
27.84 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10205410
|
Thermo Scientific Acros
122665000 |
500 g | Glass bottle |
76.18 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935051
|
Thermo Scientific Acros
122660025 |
2.5 kg | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-63-8 | |
203.02 | |
isophthaloyl chloride, isophthaloyl dichloride, 1,3-benzenedicarbonyl dichloride, m-phthaloyl chloride, isophthalic chloride, isophthalic acid chloride, isophthalic acid dichloride, isothaloyl chloride, isophthalyl chloride, 1,3-benzenedicarbonyl chloride | |
benzene-1,3-dicarbonyl chloride |
C8H4Cl2O2 | |
FDQSRULYDNDXQB-UHFFFAOYSA-N | |
7451 | |
C1=CC(=CC(=C1)C(=O)Cl)C(=O)Cl |
Specifications
99-63-8 | |
White | |
180°C | |
98% | |
C6H4(COCl)2 | |
isophthaloyl chloride, isophthaloyl dichloride, 1,3-benzenedicarbonyl dichloride, m-phthaloyl chloride, isophthalic chloride, isophthalic acid chloride, isophthalic acid dichloride, isothaloyl chloride, isophthalyl chloride, 1,3-benzenedicarbonyl chloride | |
FDQSRULYDNDXQB-UHFFFAOYSA-N | |
benzene-1,3-dicarbonyl chloride | |
7451 | |
98% | |
Isophthaloyl dichloride |
43.0°C to 45.0°C | |
276.0°C | |
Authentic | |
C8H4Cl2O2 | |
09,IV,3295 | |
Solubility in water: reacts. Other solubilities: reacts with alcohol,soluble in ether and other organic solvents | |
C1=CC(=CC(=C1)C(=O)Cl)C(=O)Cl | |
203.02 | |
203.02 | |
Crystalline Mass |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-774-7
- Isophthaloyl dichloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H312-Harmful in contact with skin.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.