missing translation for 'onlineSavingsMsg'
Learn More

Isobutyraldehyde, 99+%, Thermo Scientific Chemicals

45.53 € - 223.00 €

Chemical Identifiers

CAS 78-84-2
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00006980
InChI Key AMIMRNSIRUDHCM-UHFFFAOYSA-N
Synonym isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde
PubChem CID 6561
ChEBI CHEBI:48943
IUPAC Name 2-methylpropanal
SMILES CC(C)C=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10091130
View Documents
Thermo Scientific Acros
122510010
1 L Glass bottle
45.53 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10001140
View Documents
Thermo Scientific Acros
122510025
2.5 L Glass bottle
84.62 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10677963
View Documents
Thermo Scientific Acros
122510100
10 L Plastic drum
223.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78-84-2
72.11
AMIMRNSIRUDHCM-UHFFFAOYSA-N
6561
2-methylpropanal
C4H8O
MFCD00006980
isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde
CHEBI:48943
CC(C)C=O

Specifications

78-84-2
0.7900g/mL
−24°C
99% min. (GC)
(CH3)2CHCHO
MFCD00006980
15, 5199
isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde
AMIMRNSIRUDHCM-UHFFFAOYSA-N
2-methylpropanal
6561
0.45 mPa.s (20°C)
99+%
Isobutyraldehyde
-65.0°C
63.0°C
Authentic
C4H8O
1.3710 to 1.3740
01, 671
0.79
Solubility in water: 75g/L (20°C). Other solubilities: miscible with ethanol, ether, carbon disulfide,, acetone, benzene, toluene and chloroform
CC(C)C=O
72.11
CHEBI:48943
72.11
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond conta

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-149-6

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado