Learn More
Isobutyraldehyde, 98%, Thermo Scientific Chemicals
30.15 € - 78.99 €
Chemical Identifiers
CAS | 78-84-2 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00006980 |
InChI Key | AMIMRNSIRUDHCM-UHFFFAOYSA-N |
Synonym | isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde |
PubChem CID | 6561 |
ChEBI | CHEBI:48943 |
IUPAC Name | 2-methylpropanal |
SMILES | CC(C)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410033
|
Thermo Scientific Alfa Aesar
A12106.AP |
500 mL |
30.15 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420033
|
Thermo Scientific Alfa Aesar
A12106.0F |
2500 mL |
78.99 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isobutyraldehyde is used as an intermediate in the preparation of isobutanol, methacrolein, hydroxypivaldehyde and neopentyl glycol. It is actively involved in the Cannizaro reaction. It is also used as an intermediate to prepare pharmaceuticals, agrochemicals, vitamins, antioxidants, rubber accelerators, textile auxiliaries, perfumery and flavors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsobutyraldehyde is used as an intermediate in the preparation of isobutanol, methacrolein, hydroxypivaldehyde and neopentyl glycol. It is actively involved in the Cannizaro reaction. It is also used as an intermediate to prepare pharmaceuticals, agrochemicals, vitamins, antioxidants, rubber accelerators, textile auxiliaries, perfumery and flavors.
Solubility
Slightly miscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents.
Chemical Identifiers
78-84-2 | |
72.11 | |
AMIMRNSIRUDHCM-UHFFFAOYSA-N | |
6561 | |
2-methylpropanal |
C4H8O | |
MFCD00006980 | |
isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde | |
CHEBI:48943 | |
CC(C)C=O |
Specifications
78-84-2 | |
0.794 | |
-24°C (-11°F) | |
C4H8O | |
1.3723 | |
UN2045 | |
Air sensitive | |
isobutyraldehyde, isobutanal, isobutylaldehyde, propanal, 2-methyl, 2-methylpropionaldehyde, isobutyric aldehyde, valine aldehyde, isopropylaldehyde, 2-methyl-1-propanal, isobutaldehyde | |
AMIMRNSIRUDHCM-UHFFFAOYSA-N | |
2-methylpropanal | |
6561 | |
72.11 | |
Liquid |
°C to 65°C | |
63°C to 65°C | |
98% | |
(CH3)2CHCHO | |
MFCD00006980 | |
605330 | |
14,5154 | |
Slightly miscible with water. | |
CC(C)C=O | |
72.11 | |
CHEBI:48943 | |
98% | |
Isobutyraldehyde |
Safety and Handling
GHS H Statement
H225-H319-H335
P201-P202-P210-P233-P240-P243-P264b-P270-P280-P301+P312-P305+P351+P338-P308+P313-P330-P372-P374-P380-P501c
H225-H302-H319-H341
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYRALDEHYDE
missing translation for 'einecsNumber' : 201-149-6
missing translation for 'rtecsNumber' : NQ4025000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Air Sensitive
Recommended Storage : Keep cold; Store under Argon
- Isobutyraldehyde
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Isobutyraldehyde
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.