missing translation for 'onlineSavingsMsg'
Learn More

Isobutyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Product Code. 11410147
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18395-30-7
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.303
MDL Number MFCD00051589
InChI Key XYJRNCYWTVGEEG-UHFFFAOYSA-N
Synonym isobutyltrimethoxysilane, isobutyl trimethoxy silane, silane, trimethoxy 2-methylpropyl, trimethoxy 2-methylpropyl silane, dynasylan ibtmo, prosil 178, isobutyltrimethoxy silane, iso-butyl trimethoxysilane, acmc-1cg69, 2-methyl-1-propyltrimethoxysilane
PubChem CID 87622
IUPAC Name trimethoxy(2-methylpropyl)silane
SMILES CC(C)C[Si](OC)(OC)OC

Specifications

Boiling Point 154°C to 157°C
Quantity 500 g
UN Number UN1993
Beilstein 2233976
Flash Point 43°C (109°F)
Formula Weight 178.31
Percent Purity 97%
Refractive Index 1.396
Sensitivity Moisture sensitive
Density 0.93
Chemical Name or Material Isobutyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'