missing translation for 'onlineSavingsMsg'
Learn More

Isobutyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Product Code. 11400147
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18395-30-7
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.303
MDL Number MFCD00051589
InChI Key XYJRNCYWTVGEEG-UHFFFAOYSA-N
Synonym isobutyltrimethoxysilane, isobutyl trimethoxy silane, silane, trimethoxy 2-methylpropyl, trimethoxy 2-methylpropyl silane, dynasylan ibtmo, prosil 178, isobutyltrimethoxy silane, iso-butyl trimethoxysilane, acmc-1cg69, 2-methyl-1-propyltrimethoxysilane
PubChem CID 87622
IUPAC Name trimethoxy(2-methylpropyl)silane
SMILES CC(C)C[Si](OC)(OC)OC

Specifications

Boiling Point 154°C to 157°C
Quantity 100 g
UN Number UN1993
Beilstein 2233976
Flash Point 43°C (109°F)
Formula Weight 178.31
Percent Purity 97%
Refractive Index 1.396
Sensitivity Moisture sensitive
Density 0.93
Chemical Name or Material Isobutyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isobutyltrimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Isobutyltrimethoxysilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'