missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron Solution for ICP, 10g/L, HNO3 Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
101.00 € - 235.00 €
Specifications
Concentration | 10g/L |
---|---|
Solution Contains | HNO3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12987183
|
Fisher Chemical
S 319 100 |
100 mL |
101.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12997183
|
Fisher Chemical
S 319 250 |
250 mL |
173.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12907193
|
Fisher Chemical
S 319 500 |
500 mL |
235.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
10g/L | |
HNO3 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Iron (Fe) 10000mg/l in HNO3 5%
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title