missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Iii) Oxide, purified, Honeywell Fluka™
Purified,≥95%
38.66 € - 312.40 €
Chemical Identifiers
CAS | 1309-37-1 |
---|---|
Molecular Formula | Fe2O3 |
Molecular Weight (g/mol) | 159.69 |
MDL Number | MFCD00011008 |
InChI Key | JEIPFZHSYJVQDO-UHFFFAOYSA-N |
IUPAC Name | trioxodiiron |
SMILES | [Fe][Fe](=O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15621130
|
Honeywell-Fluka
12342-250G |
250 g |
38.66 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15611130
|
Honeywell-Fluka
12342-1KG |
1 kg |
63.25 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15641130
|
Honeywell-Fluka
12342-6X250G |
6 x 250 g |
173.99 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15631130
|
Honeywell-Fluka
12342-6X1KG |
6 x 1 kg |
312.40 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1309-37-1 | |
159.69 | |
JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
[Fe][Fe](=O)(=O)=O |
Fe2O3 | |
MFCD00011008 | |
trioxodiiron |
Specifications
1309-37-1 | |
MFCD00011008 | |
JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
trioxodiiron | |
159.69g/mol | |
Purified |
Fe2O3 | |
NONH for all modes of transport | |
[Fe][Fe](=O)(=O)=O | |
159.69 | |
≥95% | |
Iron(Iii) Oxide |
Safety and Handling
missing translation for 'einecsNumber' : 215-168-2
missing translation for 'rtecsNumber' : NO7400000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title