missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) isopropoxide, 98%, Thermo Scientific Chemicals

Product Code. 11316299
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Iron(III) isopropoxide is applied in the field of nano-materials and thin films, engineering and environmental field. It is employed in catalysis and synthesis, energy conversion and storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14995-22-3
Molecular Formula C9H21FeO3
Molecular Weight (g/mol) 233.109
MDL Number MFCD00070436
InChI Key QUHDSMAREWXWFM-UHFFFAOYSA-N
Synonym iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe
PubChem CID 11961763
IUPAC Name iron(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3]

Specifications

Color Gray
Packaging Ampule
Linear Formula Fe[OCH(CH3)2]3
Quantity 25 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 233.11
Percent Purity 98%
Physical Form Solid Mass
Chemical Name or Material Iron (III) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'