Learn More
Iron(III) hexacyanoferrate(II), Thermo Scientific Chemicals
46.40 € - 149.00 €
Chemical Identifiers
CAS | 14038-43-8 |
---|---|
Molecular Formula | C18Fe7N18 |
Molecular Weight (g/mol) | 859.239 |
MDL Number | MFCD00135663 |
InChI Key | DCYOBGZUOMKFPA-UHFFFAOYSA-N |
Synonym | prussian blue, ferric ferrocyanide, iron iii ferrocyanide, iron iii hexacyanoferrate ii, ferric hexacyanoferrate, parisian blue, berlin blue, ferrocin, preussischblau, turnbulls blau |
PubChem CID | 2724251 |
IUPAC Name | iron(2+);iron(3+);octadecacyanide |
SMILES | [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[Fe+2].[Fe+2].[Fe+2].[Fe+3].[Fe+3].[Fe+3].[Fe+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353658
|
Thermo Scientific Alfa Aesar
039259.14 |
25 g |
46.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363658
|
Thermo Scientific Alfa Aesar
039259.22 |
100 g |
149.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iron(III) hexacyanoferrate(II) is uses extensively as an Iron Oxide dye. Prussian blue is used as a paint and wallpaper printing, chemical coatings, carbon paper and in the plastics industry. as an antidote for poisoning with radioactive cesium or thallium. In the metalworking and mechanical engineering Prussian blue is thinly applied as a paste on metal surfaces in order to assess the quality scraped surfaces can.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14038-43-8 | |
859.239 | |
DCYOBGZUOMKFPA-UHFFFAOYSA-N | |
2724251 | |
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[Fe+2].[Fe+2].[Fe+2].[Fe+3].[Fe+3].[Fe+3].[Fe+3] |
C18Fe7N18 | |
MFCD00135663 | |
prussian blue, ferric ferrocyanide, iron iii ferrocyanide, iron iii hexacyanoferrate ii, ferric hexacyanoferrate, parisian blue, berlin blue, ferrocin, preussischblau, turnbulls blau | |
iron(2+);iron(3+);octadecacyanide |
Specifications
14038-43-8 | |
Powder | |
MFCD00135663 | |
It is practically insoluble in water. | |
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[Fe+2].[Fe+2].[Fe+2].[Fe+3].[Fe+3].[Fe+3].[Fe+3] | |
859.239 | |
859.25 | |
Hygroscopic |
Blue | |
C18Fe7N18 | |
prussian blue, ferric ferrocyanide, iron iii ferrocyanide, iron iii hexacyanoferrate ii, ferric hexacyanoferrate, parisian blue, berlin blue, ferrocin, preussischblau, turnbulls blau | |
DCYOBGZUOMKFPA-UHFFFAOYSA-N | |
iron(2+);iron(3+);octadecacyanide | |
2724251 | |
Odorless | |
Iron(III) hexacyanoferrate(II) |
Safety and Handling
H301-H311-H332: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled.
missing translation for 'einecsNumber' : 237-875-5
missing translation for 'rtecsNumber' : LJ8200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only